Skip to Content
Merck
All Photos(2)

Key Documents

HPA000603

Sigma-Aldrich

Anti-CRELD2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cysteine-rich with EGF-like domain protein 2 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€490.00

€490.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
100 μL
€490.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€490.00


Estimated to ship onMay 29, 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200-1:500

immunogen sequence

HLEAWWLQLKSEYPDLFEWFCVKTLKVCCSPGTYGPDCLACQGGSQRPCSGNGHCSGDGSRQGDGSCRCHMGYQGPLCTDCMDGYFSSLRNETHSICTACDESCKTCSGLTNRDCGECEVG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRELD2(79174)

General description

CRELD2 (cysteine-rich with EGF-like domains 2) maps to chromosome 22q13. It is expressed ubiquitously during development and by mature tissues. Highest expression levels are seen in adult endocrine tissues.

Immunogen

Cysteine-rich with EGF-like domain protein 2 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CRELD2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CRELD2 (cysteine-rich with EGF-like domains 2) interacts with the large cytoplasmic domain of α4 and β2 subunits of human neuronal nicotinic acetylcholine receptor and may regulate their expression and transport. It is an ER stress-inducible factor localized to the ER-Golgi apparatus. It is involved in BMP (bone morphogenetic protein) 9-regulated osteogenic differentiation of mesenchymal stem cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73355

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiye Zhang et al.
PloS one, 8(9), e73086-e73086 (2013-09-11)
Mesenchymal stem cells (MSCs) are multipotent progenitors that can undergo osteogenic differentiation under proper stimuli. We demonstrated that BMP9 is one of the most osteogenic BMPs. However, the molecular mechanism underlying BMP9-initiated osteogenic signaling in MSCs remains unclear. Through gene
Cheryl L Maslen et al.
Gene, 382, 111-120 (2006-08-22)
CRELD2 is the second member of the CRELD family of proteins. The only other CRELD family member, encoded by CRELD1, is also known as the AVSD2 gene as mutations in CRELD1 are associated with cardiac atrioventricular septal defects (AVSD). Like
José A Ortiz et al.
Journal of neurochemistry, 95(6), 1585-1596 (2005-10-22)
Using a yeast two-hybrid screening we report the isolation of a novel human protein, hCRELD2beta, that interacts specifically with the large cytoplasmic regions of human nicotinic acetylcholine receptor (nAChR) alpha4 and beta2 subunits, both in yeast cells and in vitro.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service