Skip to Content
Merck
All Photos(1)

Key Documents

H7149

SAFC

(±)-6-Hydroxy-3,4-methylenedioxymethamphetamine

Synonym(s):

6-Hydroxy-MDMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO3
CAS Number:
Molecular Weight:
209.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage temp.

2-8°C

SMILES string

CNC(C)Cc1cc2OCOc2cc1O

InChI

1S/C11H15NO3/c1-7(12-2)3-8-4-10-11(5-9(8)13)15-6-14-10/h4-5,7,12-13H,3,6H2,1-2H3

InChI key

RQQIIXFYTHWFKW-UHFFFAOYSA-N

Biochem/physiol Actions

A metabolite of MDMA

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lim, H.K. and Foltz, R.L.
Biomedical Mass Spectrometry, 20, 677-677 (1991)
I Elayan et al.
European journal of pharmacology, 221(2-3), 281-288 (1992-10-20)
The long-term effects of three metabolites of 3,4-methylenedioxymethamphetamine (MDMA) on the central monoaminergic systems of the rat were examined. Seven days after the intracerebroventricular administration of 0.25 and 0.5 mumol 2,4,5-trihydroxyamphetamine, hippocampal tryptophan hydroxylase (TPH) activity was reduced to 5
H K Lim et al.
Biological mass spectrometry, 20(11), 677-686 (1991-11-01)
Aromatic hydroxylation has been established as a pathway for the in vivo metabolism of 3,4-(methylenedioxy)methamphetamine (MDMA) in the rat. Hydroxylation occurred at positions 2, 5 and 6 of the 3,4-methylenedioxyphenyl ring, but is favored at the 6 position. All three
T Chu et al.
Biochemical pharmacology, 51(6), 789-796 (1996-03-22)
3,4-Methylenedioxymethamphetamine (MDMA) affects both dopamine and serotonin (5-HT) systems. One of its acute actions is to cause a reversible fall in steady-state brain 5-HT concentrations. To investigate the chemical basis of this acute effect, the brain levels of the parent
T M Walker et al.
Cell biology and toxicology, 15(3), 137-142 (1999-12-02)
Rat whole-brain spheroids were used to assess the intrinsic neurotoxicity of methylenedioxy-methamphetamine (MDMA, Ecstasy) and two of its metabolites, dihydroxymethamphetamine (DHMA) and 6-hydroxy-MDMA (6-OH MDMA). Exposure of brain spheroids to MDMA or the metabolite 6-OH MDMA (up to 500 micromol/L)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service