Skip to Content
Merck
All Photos(1)

Key Documents

H6263

Sigma-Aldrich

Hydra Peptide Fragment 7-11

≥97% (HPLC)

Synonym(s):

Lys-Val-Ile-Leu-Phe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H54N6O6
CAS Number:
Molecular Weight:
618.81
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(NC(=O)C(N)CCCCN)C(C)C)C(=O)NC(CC(C)C)C(=O)NC(Cc1ccccc1)C(O)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H C Schaller et al.
European journal of biochemistry, 138(2), 365-371 (1984-01-16)
By exposing different sites of the 'head activator', different sets of antibodies were designed and produced which recognised either the amino or the carboxy terminus of the free 'head activator', which reacted with 'head activator' in a tissue-fixed conformation, or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service