Skip to Content
Merck
All Photos(1)

Documents

GW21285B

Sigma-Aldrich

Anti-AXO antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Axotrophin variant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21285B

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MARCH7(64844)

Immunogen

Immunogen Sequence: GI # 12383066, sequence 481-680
Recombinant axotrophin

Application

Anti-AXO antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

E3 ubiquitin-protein ligase MARCH7 (axotrophin) is an enzyme encoded by the MARCH7 gene in humans and belongs to the MARCH family of proteins. It is a novel family of transmembrane ubiquitin ligases that target glycoproteins for lysosomal destruction via ubiquitination of the cytoplasmic tail. MARCH-7 was first identified in mouse embryonic stem cells as a neural stem cell gene. It has a distinct expression profile from that of other MARCH family members.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Bartee et al.
Journal of virology, 78(3), 1109-1120 (2004-01-15)
Poxviruses and gamma-2 herpesviruses share the K3 family of viral immune evasion proteins that inhibit the surface expression of glycoproteins such as major histocompatibility complex class I (MHC-I), B7.2, ICAM-1, and CD95(Fas). K3 family proteins contain an amino-terminal PHD/LAP or
Cristina A Szigyarto et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 58(4), 301-308 (2009-11-11)
Axotrophin/MARCH-7 was first identified in mouse embryonic stem cells as a neural stem cell gene. Using the axotrophin/MARCH-7 null mouse, we discovered profound effects on T lymphocyte responses, including 8-fold hyperproliferation and 5-fold excess release of the stem cell cytokine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service