GEPA63131
CY®3B Maleimide 1mg
Synonym(s):
CY3B Maleimide, Maleimide Dye
About This Item
Recommended Products
manufacturer/tradename
Cytiva
General description
Features and Benefits
- Store refrigerated at 2–8°C in the dark.
- Reconstitute the material to 1 mg/mL in anhydrous DMF/DMSO. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
- Produces an intense signal easily detected using most rhodamine fi lter sets in the appropriate instrumentation.
Storage and Stability
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service