Skip to Content
Merck
All Photos(1)

Key Documents

GE18672700

Sample pump P960

ÄKTA® Systems, Cytiva 18672700

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115705
NACRES:
NB.13
Pricing and availability is not currently available.

AC/DC input

32 V DC

description

The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100% Ethyl acetate, 100% Hexane and 100% Tetrahydrofuran.

feature

Connections: male M6 (pump inlet) male 1/16” finger-tight (pump outlet) male 1/16” finger-tight (pressure sensor inlet) male 1/16” finger-tight pressure sensor outlet
Relative Humidity: 20-95% (non condensing)
Wetted Materials" Ethylenetetrafluoroethylene (ETFE) Polyetheretherketone (PEEK) Polytetrafluoroethylene (PTFE) Perfluor rubber (PFR) Ultra high molecular weight polyethylene (UHMWPE) Titanium alloy (Ti alloy) Borosilicate glass Zirconia Cermaic Elgiloy
ÄKTA® Systems

manufacturer/tradename

Cytiva 18672700

parameter

20 bar max. pressure
40 °C max. temp.
50 mL/min max. flow rate

depth × width

(99 mm) (148 mm) (158 mm)

Related Categories

Legal Information

ÄKTA is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service