Skip to Content
Merck
All Photos(1)

Key Documents

GE18-1153-17

Sigma-Aldrich

TRICORN 10/200 GLASS COLUMN

Cytiva 18-1153-17, pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
SB.52

material

glass column

form

prepacked column

feature

wetted part

packaging

pack of 1 ea

manufacturer/tradename

Cytiva 18-1153-17

technique(s)

liquid chromatography (LC): suitable

General description

Glass tube 10/200

Features and Benefits

  • Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures (see Specifications). They are suitable for use with high performance media, such as Sepharose High Performance, Superdex Prep Grade, and SOURCE.
  • The columns are available with inner diameters of 5 and 10 mm, and in lengths from 20 to 200 mm. Column lengths of 300 and 600 mm are also available for the 10 mm diameter columns. The design and dimensions of the columns make them particularly useful for screening and polishing steps in protein purification.
  • Glass tubes coated with plastic film protects individuals in the laboratory from harm if the column is dropped or operated outside its specification.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service