Skip to Content
Merck
All Photos(2)

Documents

GBL204871

Grace Bio-Labs ProPlate® tray and cover

for Delrin® snap clip modules, holds 4 slide modules

Synonym(s):

slide holder, slide module

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.15

sterility

non-sterile

packaging

pack of 1 ea

manufacturer/tradename

Grace Bio-Labs 204871

external size

85.3 mm × 127.5 mm

color

none

Looking for similar products? Visit Product Comparison Guide

General description

Organize up to 4 chambered slides into the footprint of a microtiter plate for use with automated liquid handling robots or plate washers.

Legal Information

Delrin is a registered trademark of E. I. du Pont de Nemours and Company
ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service