Skip to Content
Merck
All Photos(1)

Documents

G8770

Sigma-Aldrich

Growth Hormone Releasing Factor Fragment 1-40 human

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C194H317N61O63S
CAS Number:
Molecular Weight:
4544.03
MDL number:
UNSPSC Code:
12352202

Assay

≥90% (HPLC)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... GHRH(2691)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Ala-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-Gln-Gln-Gly-Glu-Ser-Asn-Gln-Glu-Arg-Gly-Ala

Biochem/physiol Actions

Equipotent to GRF (1-44) in stimulating release of somatotropin from anterior pituitary; endogenous peptide originally found in human pancreatic tumor cells, but also seen in hypothalamus

Linkage

Similar to G 0263, but prepared by Sigma.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service