G4144
Gly-Arg-Ala-Asp-Ser-Pro-Lys
≥97% (HPLC)
Synonym(s):
GRADSPK
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C29H51N11O11
CAS Number:
Molecular Weight:
729.78
MDL number:
UNSPSC Code:
12352200
Assay
≥97% (HPLC)
composition
Peptide content, ~60%
storage temp.
−20°C
SMILES string
C[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)CN)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CO)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CCCCN)C(O)=O
Biochem/physiol Actions
Negative control peptide for the integrin-binding peptide GRGDSPK.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Clément Faye et al.
The Journal of biological chemistry, 284(33), 22029-22040 (2009-06-09)
Endostatin is an endogenous inhibitor of angiogenesis. Although several endothelial cell surface molecules have been reported to interact with endostatin, its molecular mechanism of action is not fully elucidated. We used surface plasmon resonance assays to characterize interactions between endostatin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service