Skip to Content
Merck
All Photos(1)

Key Documents

G4127

Sigma-Aldrich

Gly-Val

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O3
CAS Number:
Molecular Weight:
174.20
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)CN)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N A Basova et al.
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova, 93(12), 1423-1434 (2008-03-06)
Energy-dependent accumulation of glycine and glycyl-L-valine within the small intestinal mucosa in a chicken model of in vitro local oxygenation of the small intestinal preparation was studied. It has been shown that the most effective bilateral oxygenation significantly increase accumulation
In vivo and vitro absorption of amino acids and dipeptides in the small intestine of uremic rats.
G Sterner et al.
Nephron, 31(3), 273-276 (1982-01-01)
Carl Henrik Görbitz et al.
Acta crystallographica. Section C, Crystal structure communications, 63(Pt 1), o58-o60 (2007-01-09)
The title peptide, C7H14N2O3, crystallizes with seven independent molecules in the asymmetric unit. All have essentially the same overall conformation, but some flexibility is exhibited by the glycine residue. It appears that the high Z' value, observed only three times
I L Tarvid et al.
Zhurnal evoliutsionnoi biokhimii i fiziologii, 19(6), 550-555 (1983-11-01)
Studies have been made of changes in catalytic activity and regulatory properties of membrane and intracellular glycyl-valine dipeptidase from the small intestine of chicks ageing from 1 to 40 days. Gradual decrease in the activity of membrane glycyl-valine dipeptidase was
S Janarthanan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 105, 34-37 (2013-01-08)
A nonlinear optical crystal of N-Glycyl-L-Valine (GV) single crystals was grown by slow evaporation solution growth technique from an aqueous solution. The unit cell parameters and the crystal structure were determined by single crystal X-ray diffraction study. The Fourier transform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service