Recommended Products
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The American journal of physiology, 251(5 Pt 1), G636-G641 (1986-11-01)
Quantitative autoradiography was used to measure the distribution of transport of leucine, lysine, and glycyl-L-leucine along the villus of the rat jejunum. A comparison was made between normal juvenile animals and ones maintained on a 5% protein isocalorific diet. Uptake
Biochemical pharmacology, 45(3), 776-779 (1993-02-09)
The human colon adenocarcinoma cell line Caco-2 retains the H+/dipeptide cotransporter. To identify the structure of the human dipeptide transporter, we have examined the expression of the transporter in Xenopus laevis oocytes injected with Caco-2 poly(A)+RNA, by monitoring the uptake
The American journal of physiology, 254(5 Pt 1), E588-E594 (1988-05-01)
Oligopeptides may enter the bloodstream from endogenous and exogenous sources. Using an organ-balance technique in conscious dogs, we investigated the role of individual organs in removal of two model oligopeptides (glycylleucine and glycylglycine) from plasma under steady-state conditions. Despite an
Journal of biomaterials science. Polymer edition, 21(6-7), 843-862 (2010-05-21)
The inherent flexibility of polyurethane (PU) chemistry allows the incorporation of specific chemical moieties into the backbone structure conferring a unique biological function to these synthetic polymers. We describe here the synthesis and characterization of a PU containing a Gly-Leu
Bioscience, biotechnology, and biochemistry, 76(1), 125-128 (2012-01-11)
Two dipeptides, glycyl-L-leucine (G-L) and L-leucyl-glycine (L-G), the concentrations of which were 10 mmol/L, were degraded in subcritical water in order to understand fully the phenomena occurring during treatment. Treatment was administered in a stainless steel tubular reactor, which was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service