Skip to Content
Merck
All Photos(1)

Key Documents

F5513

Sigma-Aldrich

Carestream® BioMax® XAR Film

Synonym(s):

autoradiography film, xray film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105341
NACRES:
NB.22

size

8 in. (20 cm) × 10 in. (25 cm)

Looking for similar products? Visit Product Comparison Guide

General description

  • Coated with emulsion on both sides of the clear base.
  • Recommended for chemiluminescence.
  • Very versatile for autoradiography with high or low energy isotopes.
  • Sensitivity can be increased with intensifying screens when using low-energy isotopes.
  • When handling film in a darkroom, Kodak LED Safelight Darkroom Illuminator is recommended.
  • All the XAR films are the same, just packaged differently.
Bulk packed

Footnote

Kodak X-OMAT AR film was reformulated to become BioMax XAR film.

Legal Information

BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yibo Xue et al.
Nature communications, 10(1), 558-558 (2019-02-06)
Inactivating mutations in SMARCA4 (BRG1), a key SWI/SNF chromatin remodelling gene, underlie small cell carcinoma of the ovary, hypercalcemic type (SCCOHT). To reveal its druggable vulnerabilities, we perform kinase-focused RNAi screens and uncover that SMARCA4-deficient SCCOHT cells are highly sensitive
Eri Takeda et al.
PloS one, 6(11), e27815-e27815 (2011-11-24)
Vpr, an accessory protein of human immunodeficiency virus type 1, is a multifunctional protein that plays an important role in viral replication. We have previously shown that the region between residues 17 and 74 of Vpr (Vpr(N17C74)) contained a bona
H F Zhao et al.
Neuroscience, 310, 641-649 (2015-10-11)
Our previous studies demonstrated resveratrol (Res) administration protected Alzheimer's disease (AD) rats from developing memory decline by anti-oxidation. Beta-amyloid peptide 1-42 (Aβ1-42) is not only the primary protein component of senile plaques in AD but also is believed to play

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service