Skip to Content
Merck
All Photos(1)

Key Documents

F107

Sigma-Aldrich

Flunarizine dihydrochloride

Synonym(s):

1-[Bis(4-fluorophenyl)methyl]-4-(3-phenyl-2-propenyl)piperazine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26F2N2 · 2HCl
CAS Number:
Molecular Weight:
477.42
Beilstein:
4284243
EC Number:
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

SMILES string

Cl[H].Cl[H].Fc1ccc(cc1)C(N2CCN(CC2)C\C=C\c3ccccc3)c4ccc(F)cc4

InChI

1S/C26H26F2N2.2ClH/c27-24-12-8-22(9-13-24)26(23-10-14-25(28)15-11-23)30-19-17-29(18-20-30)16-4-7-21-5-2-1-3-6-21;;/h1-15,26H,16-20H2;2*1H/b7-4+;;

InChI key

RXKMOPXNWTYEHI-RDRKJGRWSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks T-type Ca2+/Na+ channels.
Blocks T-type Ca2+/Na+ channels; inhibits K+-induced catecholamine release from chromaffin cells

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin-Peng Wang et al.
Pain, 152(8), 1864-1871 (2011-05-28)
Insufficient clinical trial data were available to prove the efficacy of acupuncture for migraine prophylaxis. A multicenter, double-dummy, single-blinded, randomized controlled clinical trial was conducted at the outpatient departments of acupuncture at 5 hospitals in China to evaluate the effectiveness
Carlijn El Hoekstra et al.
The Cochrane database of systematic reviews, (7)(7), CD007960-CD007960 (2011-07-08)
Tinnitus is the perception of sound or noise in the absence of an external or internal acoustic stimulation. It is a common and potentially distressing symptom for which no adequate therapy exists. To assess the effectiveness of anticonvulsants in patients
A Van Driessche et al.
Acta clinica Belgica, 67(2), 123-126 (2012-06-21)
Cyclic vomiting syndrome (CVS) is a functional disorder that is considered to be a manifestation of migraine diathesis. It is characterized by stereotypical episodes of severe nausea and vomiting lasting several hours or days, with return to baseline health between
Ning Luo et al.
Pain medicine (Malden, Mass.), 13(1), 80-86 (2012-01-12)
The objective of this study was to observe the efficacy, safety, and side effects of a combination of flunarizine plus topiramate compared with either flunarizine and or toparamate alone for migraine prophylaxis. Out of 150 patients with migraine recruited into
Barbara Budzynska et al.
Behavioural brain research, 228(1), 144-150 (2011-12-20)
The motivational component of drug withdrawal may contribute to drug seeking and relapse through the negative reinforcement-related process; thus, it is important to understand the mechanisms that mediate affective withdrawal behaviors. The present study was undertaken to examine the calcium-dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service