Skip to Content
Merck
All Photos(1)

Key Documents

F0639

Sigma-Aldrich

Fmoc-L-aspartic acid (β-tert-butyl) Sasrin® resin ester

200-400 mesh

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

extent of labeling

0.3-0.8 mmol FMOC-amino acid per g resin

particle size

200-400 mesh

functional group

Fmoc

storage temp.

2-8°C

Application

For peptide synthesis. Peptides can be cleaved from the resin with 0.5-1% CF3CO2H in CH2Cl2.

Other Notes

N-FMOC-amino acid ester of 3-methoxy-4-oxymethyl-phenoxymethylated 1% divinylbenzene cross-linked polystyrene.

Legal Information

Sasrin is a registered trademark of Bachem Bioscience Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service