Skip to Content
Merck
All Photos(1)

Documents

EPF740540035

Eppendorf® CryoCube® F740h

capacity 740 L, cooling, 5 Classic interface, AC/DC input 120 - 208 V, 60 Hz

Synonym(s):

Eppendorf® CryoCube® F740h, CryoCube® F740h

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103034

feature

cooling 5 Classic interface

capacity

740 L

Looking for similar products? Visit Product Comparison Guide

General description

The Eppendorf CryoCube F740 series is one of the newest additions to the Eppendorf ultra-low temperature freezer family (also called ultra-low temperature freezer, -80°C). The CryoCube F740 series of ultra-low temperature freezers combines the traditionally high quality of our previous ultra-low temperature freezers with a higher capacity to safely store more of your samples while optimizing energy consumption. By incorporating the PhysioCare Concept into the development, the CryoCube F740 is lighter and more comfortable to use, supports ergonomics in the workplace and enables an optimal workflow in the laboratory.

Application

Cryopreservation, Cryostorage

Legal Information

CryoCube is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service