EP5702712000
Eppendorf® Adapter for 90 mL rectangular bucket for use with standard tubes
Holds 15 mL tubes, pack of 2 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
Holds 15 mL tubes
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702712000
technique(s)
centrifugation: suitable
compatibility
for use with 90 mL rectangular buckets
Looking for similar products? Visit Product Comparison Guide
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service