EP0030702018
Eppendorf® Cell Culture Dishes
non-treated surface, size 100 mm, case of 300 ea (Internal packs of 10)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.13
material
polystyrene
sterility
sterile
feature
lid
packaging
case of 300 ea (Internal packs of 10)
manufacturer/tradename
Eppendorf® 0030702018
diam. × H
93.8 mm × 19.4 mm
dish capacity
99.2 mL
dish diam.
93.8 mm
maximum volume
99.2 mL
size
100 mm
surface area
21.9 cm2
working volume
8.0-10.0 mL
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Whenever you need direct access to your cells, dishes are the format of choice. Eppendorf cell culture dishes were designed with a focus on improved handling and stacking performance to ensure a new level of safe and carefree usage. It all starts with the packaging: the innovative design can be easily opened without tools and resealed without tape. The bags are shrinkable for space-saving, safe, and contamination-free storage of unused dishes. With the Eppendorf dishes, a corrugated handling ring provides unsurpassed safety in handling during transportation and expansion of cells. A SplashProtect ring inside the dish lid prevents spillage and traps fluid if condensation occurs. Combined with the outstanding, sturdy stacking, our cell culture dishes are unparalleled in their ease of transport and handling, safety, and convenience.
Innovative packaging
The packaging combines tool-free opening, resealable top and side for tape-free closing and an easy solution to shrink the package for space-conscious storage
Safety in all situations
A corrugated handling ring provides unsurpassed safety in handling during transportation and while working with dishes. The SplashProtect ring inside the dish lid traps liquid and prevents spills during transportation or incubation.
Direct surface identification
Easy surface reference and robust stacking performance by pronounced rims on dish lid and exact fitting of lid and bottom
Innovative packaging
The packaging combines tool-free opening, resealable top and side for tape-free closing and an easy solution to shrink the package for space-conscious storage
Safety in all situations
A corrugated handling ring provides unsurpassed safety in handling during transportation and while working with dishes. The SplashProtect ring inside the dish lid traps liquid and prevents spills during transportation or incubation.
Direct surface identification
Easy surface reference and robust stacking performance by pronounced rims on dish lid and exact fitting of lid and bottom
Application
Expansion and culture of adherent and suspension cells on TC treated or on non-treated polystyrene
Performance of cell-based assays and microscopic analysis
Performance of cell-based assays and microscopic analysis
Features and Benefits
Unsurpassed safety in handling during transportation and while working due to corrugated handling ring
SplashProtect ring inside of the dish lid traps liquid and prevents spills during transportation or incubation
Pronounced rim on dish lid and exact fitting of lid and bottom ensure sturdy, secure stacking
Easy differentiation of lid and dish bottom prevents unintended removal of the lid
Packaging combines tool-free opening, resealable top and side for tape-free closing, and an easy solution to shrink the package for space-conscious storage
SplashProtect ring inside of the dish lid traps liquid and prevents spills during transportation or incubation
Pronounced rim on dish lid and exact fitting of lid and bottom ensure sturdy, secure stacking
Easy differentiation of lid and dish bottom prevents unintended removal of the lid
Packaging combines tool-free opening, resealable top and side for tape-free closing, and an easy solution to shrink the package for space-conscious storage
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service