Skip to Content
Merck
All Photos(1)

Key Documents

EP0030523105

Eppendorf® Deepwell plates, DNA LoBind, 384 wells

white plate, conical bottom colorless wells, capacity 200 μL, pkg of 40 ea

Sign Into View Organizational & Contract Pricing

Select a Size

40 EA
€490.00

€490.00


Estimated to ship onApril 23, 2025



Select a Size

Change View
40 EA
€490.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
conical bottom colorless wells
white plate
capacity:
200 μL
size:
384 wells
working volume:
200 μL

€490.00


Estimated to ship onApril 23, 2025


material

conical bottom colorless wells
white plate

packaging

pkg of 40 ea

manufacturer/tradename

Eppendorf® 0030523105

technique(s)

PCR: suitable

capacity

200 μL

size

384 wells

working volume

200 μL

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf′s innovative DNA LoBind technology reduces sample-to-surface binding to ensure maximum nucleic acid recovery and offer the ideal solution for sample preparations and long-term storage. A combination of special manufacturing technologies and selected polypropylene batches ensure nearly 100 % recovery of DNA/RNA molecules-without surface coating to eliminate the risk of sample contamination. This makes Eppendorf LoBind Plates ideal for applications where sample recovery is key - such as the preparation of long-term storage nucleic acids in forensic, microarrays and NGS applications.

Application

  • Preparation or storage of DNA and RNA samples
  • Forensic trace analysis
  • Preparing dilution series in quantitative qPCR
  • Sample preparation in next-generation sequencing
  • Creation of genomic or oligonucleotide libraries

Features and Benefits

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • High-contrast Unique OptiTrack matrix: up to 30 % faster sample identification and fewer pipetting errors
  • RecoverMax well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service