Skip to Content
Merck
All Photos(1)

Key Documents

EHU041011

Sigma-Aldrich

MISSION® esiRNA

targeting human HPSE

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€195.00
50 μG
€345.00

€195.00


Estimated to ship onMay 23, 2025



Select a Size

Change View
20 μG
€195.00
50 μG
€345.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€195.00


Estimated to ship onMay 23, 2025


description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTGCAGCTGGCTTTATGTGGCTGGATAAATTGGGCCTGTCAGCCCGAATGGGAATAGAAGTGGTGATGAGGCAAGTATTCTTTGGAGCAGGAAACTACCATTTAGTGGATGAAAACTTCGATCCTTTACCTGATTATTGGCTATCTCTTCTGTTCAAGAAATTGGTGGGCACCAAGGTGTTAATGGCAAGCGTGCAAGGTTCAAAGAGAAGGAAGCTTCGAGTATACCTTCATTGCACAAACACTGACAATCCAAGGTATAAAGAAGGAGATTTAACTCTGTATGCCATAAACCTCCATAATGTCACCAAGTACTTGCGGTTACCCTATCCTTTTTCTAACAAGCAAGTGGATAAATACCTTCTAAGACCTTTGGGACCTCATGGATTACTTTCCAAATCTGTCCAACTCAATGGTCTAACTCTAAAGATGGTGGATGATCAAACCTTGCCACCTTTAATGGAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marjolein Garsen et al.
The American journal of pathology, 186(4), 805-815 (2016-02-14)
Heparanase, a heparan sulfate (HS)--specific endoglucuronidase, mediates the onset of proteinuria and renal damage during experimental diabetic nephropathy. Glomerular heparanase expression is increased in most proteinuric diseases. Herein, we evaluated the role of heparanase in two models of experimental glomerulonephritis
Haiyan Zheng et al.
Cancer biomarkers : section A of Disease markers, 15(5), 525-534 (2015-01-15)
Heparanase(HPSE), an endo-β -D-glucuronidase, is found overexpressed in Ovarian cancer (OC). The purpose of our work was to investigate primitively the possible role of HPSE in the development of OC. In this study, RNA interference (RNAi) with a HPSE small
Uri Barash et al.
International journal of cancer, 145(6), 1596-1608 (2019-04-30)
Heparanase is an endo-β-d-glucuronidase that cleaves heparan sulfate (HS) side chains of heparan sulfate proteoglycans. Compelling evidence tie heparanase levels with all steps of tumor formation including tumor initiation, growth, metastasis and chemo-resistance, likely involving augmentation of signaling pathways and
Jianping Li et al.
American journal of cancer research, 7(2), 234-244 (2017-03-25)
Heparanase (HPSE1) is elevated in various types of cancers including cervical cancer, and correlated with poor prognosis. Current study is to investigate the effects of HPSE1 on radiation response in cervical cancer. Colony formation assays after radiation were performed to
Satvik R Hadigal et al.
Nature communications, 6, 6985-6985 (2015-04-29)
Herpesviruses exemplified by herpes simplex virus-1 (HSV-1) attach to cell surface heparan sulfate (HS) for entry into host cells. However, during a productive infection, the HS moieties on parent cells can trap newly exiting viral progenies and inhibit their release.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service