Skip to Content
Merck
All Photos(1)

Key Documents

EHU017331

Sigma-Aldrich

MISSION® esiRNA

targeting human TIMP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGCGGTCAGTGAGAAGGAAGTGGACTCTGGAAACGACATTTATGGCAACCCTATCAAGAGGATCCAGTATGAGATCAAGCAGATAAAGATGTTCAAAGGGCCTGAGAAGGATATAGAGTTTATCTACACGGCCCCCTCCTCGGCAGTGTGTGGGGTCTCGCTGGACGTTGGAGGAAAGAAGGAATATCTCATTGCAGGAAAGGCCGAGGGGGACGGCAAGATGCACATCACCCTCTGTGACTTCATCGTGCCCTGGGACACCCTGAGCACCACCCAGAAGAAGAGCCTGAACCACAGGTACCAGATGGGCTGCGAGTGCAAGATCACGCGCTGCCCCATGATCCCGTGCTACATCTCCTCCCCGGACGAGTGCCTCTGGATGGACTGGGTCACAGAGAAGAACATCAACGGGCACCAGGCCAAGTTCTTCGCCTGCATCAAGAGAAGTGACGGCTCCTGTGCGTGGTACCGCGGCGCGGCGCCCCCCAAGCAGGAGTTTCTCGAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guijun Zhang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 118, 109309-109309 (2019-09-24)
To explore the roles of long noncoding RNA (lncRNA) FOXF1 Adjacent Non-Coding Developmental Regulatory RNA (FENDRR) in human non-small cell lung cancer (NSCLC). The levels of FENDRR in NSCLC cells and tissues were analyzed using qRT-PCR assay. The growth and
Shanlan Yin et al.
Experimental and therapeutic medicine, 17(4), 2837-2846 (2019-03-25)
Human papillomaviruses (HPVs) have important roles in the development and progression of cervical cancer, but the underlying mechanisms are yet to be fully elucidated. MicroRNA-130a (miR-130a) has previously been reported to promote cervical cancer growth. However, the underlying molecular mechanisms
Ji-Xiang Cao
Oncology reports, 41(6), 3367-3376 (2019-04-20)
Aberrantly expressed miRNAs play a crucial role in the progression of lung adenocarcinoma. However, to date, the role of miR‑888 in lung adenocarcinoma progression is unclear. In the present study, the biological function of miR‑888 and its underlying mechanism in
Yinghua Guo et al.
The international journal of biochemistry & cell biology, 99, 203-210 (2018-04-21)
Radiotherapy is a widely used effective treatment for lung cancer in clinic. MicroRNAs (miRNAs) have been proved to play an important role in radiation response. This study aimed to explore the regulatory role and mechanism of miR-373 in lung cancer
Hao Guan et al.
PloS one, 12(12), e0189490-e0189490 (2017-12-09)
MicroRNAs (miRNAs) are important regulators of pathobiological processes in various cancer. In the present study, we demonstrated that miR-93 expression was significantly up-regulated in gastric cancer tissues compared with that in matched normal mucosal tissues. High expression of miR-93 was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service