Skip to Content
Merck
All Photos(1)

Key Documents

E6265

Sigma-Aldrich

trans-11-Eicosenoic acid

≥99% (capillary GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H38O2
CAS Number:
Molecular Weight:
310.51
Beilstein:
1727314
MDL number:
UNSPSC Code:
12352211

Assay

≥99% (capillary GC)

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCCCC(O)=O

InChI

1S/C20H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h9-10H,2-8,11-19H2,1H3,(H,21,22)/b10-9+

InChI key

BITHHVVYSMSWAG-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service