Skip to Content
Merck
All Photos(1)

Key Documents

CLS2595

Corning® 96 Well Clear PVC Assay Microplate

flat bottom clear, PVC, bag of 25 ×, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
PVC
flat bottom clear
round wells
size:
96 wells
Pricing and availability is not currently available.

material

PVC
flat bottom clear
round wells

sterility

non-sterile

feature

lid: no

manufacturer/tradename

Corning 2595

packaging

bag of 25 ×

size

96 wells

well volume

250 μL

well working volume

50-150 μL

color

clear

suitability

suitable for (cell cuture)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Clear PVC Assay Microplate is economical and not treated.
  • Applicable for solution-based assays, serial dilutions, and general storage purposes.
  • Lids are not available.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service