Skip to Content
Merck
All Photos(1)

Documents

MH01

Reactivity: Anti-Neurones, 93062401

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Human x Mouse Hybridoma

ImmunoDon

Human circulating B lymphocytes

Myeloma

NS0

Reactivity

Anti-neurones

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Not specified

Cell Line Description

Human circulating B lymphocytes transformed with EBV. MH01 was derived from cell line 154 described in the reference given. No exogenous immunogen applied, antibody production was spontaneous.

Immunogen

None

Application

The isotype of the monoclonal antibody produced by this cell line is human IgM.

Culture Medium

RPMI 1640 + 2 mM glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service