THG3-MP-2-19-3-8
Reactivity: Anti-NP (4-hydroxy-3 nitrophenacetyl), 88012701
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
Mouse + Human Transfectoma
ImmunoDon
Not applicable
Myeloma
J558L
Reactivity
Anti-NP (4-hydroxy-3 nitrophenacetyl)
technique(s)
cell culture | mammalian: suitable
isotype
IgG3 G3m(b)
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse J558L + Mouse VH gene + Human C(gamma 3) gene
Cell Line Description
This human chimeric antibody is specifically directed against the hapten-4-hydroxy-3-nitrophenacetyl. V lambda of the endogenous light chain of plasmacytoma line complements VNP of transfected heavy chain to yield specific antibody.
Culture Medium
DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service