Skip to Content
Merck
All Photos(1)

Key Documents

AC2

86061902, mouse blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

AC2, 86061902

biological source

mouse blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Interleukin 3 (IL-3)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse lymphoblast

Cell Line Description

Derived from a mouse basophil clone and is dependent on Interleukin-3 (IL-3) for growth. IL-3 may be added in the form of Wehi conditioned media (10-20%). Culture densities between 10,000 and 100,000 cells/ml is necessary to retain cells in exponential growth phase.

Application

Growth factor studies, IL-2 assays

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 10,000 to 100,000 cells/ml; 5% CO2; 37°C. Cells will rapidly die at high densities above 5x100,000 cells/ml. Culture very sensitive to agitation.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service