HRA-19
10012802, human colon, Epithelial
Synonym(s):
HRA, HRA19
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
HRA-19, 10012802, from human colon
biological source
human colon
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
Cell Line Origin
Human colorectal adenocarcinoma
DNA Profile
STR-PCR Data:
Amelogenin: X,Y
CSF1PO: 12
D13S317: 11,12
D16S539: 11
D5S818: 11
D7S820: 9,12
THO1: 8
TPOX: 11
vWA: 16
Amelogenin: X,Y
CSF1PO: 12
D13S317: 11,12
D16S539: 11
D5S818: 11
D7S820: 9,12
THO1: 8
TPOX: 11
vWA: 16
Culture Medium
DMEM (High glucose Sigma D6546) + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:5 to 1:10 weekly using 0.05% trypsin/EDTA; 7.5% CO2; 37 °C.Suggested seeding density 2-4x10,000 cells/cm2.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service