Documents
BAF369310001
Bel-Art® Earth Friendly 1001 Stick
case of 500
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
material
( PLA (polylactic acid) resin )
packaging
case of 500
manufacturer/tradename
Bel-Art F36931-0001
L × W × H
5 3/4 in. × 3/4 in. × 1/16 in.
General description
Lessen environmental impact with these PLA (polylactic acid) resin sampling sticks. The production of 100% plant-based PLA contributes less greenhouse gasses and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Whether you are mixing samples, handling capsules, scraping containers or positioning solid samples for examination, 1001 Sticks are ready to assist! Use a separate stick for each material sampled to avoid cross contamination.
Features and Benefits
Green alternative to petroleum-based sampling tools; helps meet sustainability goals
Comparable in strength and durability to polystyrene, polypropylene and other petroleum-based plastic resin sampling tools
Disposal options include industrial composting for biodegradability, incineration, recycling and landfill
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP