Skip to Content
Merck
All Photos(1)

Documents

B9760

Sigma-Aldrich

Boc-Ile-Glu-Gly-Arg-7-amido-4-methylcoumarin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H50N8O10 · HCl
CAS Number:
Molecular Weight:
767.27
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Cl.CCC(C)C(NC(=O)OC(C)(C)C)C(=O)NC(CCC(O)=O)C(=O)NCC(=O)NC(CCCNC(N)=N)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C34H50N8O10.ClH/c1-7-18(2)28(42-33(50)52-34(4,5)6)31(49)41-23(12-13-26(44)45)29(47)38-17-25(43)40-22(9-8-14-37-32(35)36)30(48)39-20-10-11-21-19(3)15-27(46)51-24(21)16-20;/h10-11,15-16,18,22-23,28H,7-9,12-14,17H2,1-6H3,(H,38,47)(H,39,48)(H,40,43)(H,41,49)(H,42,50)(H,44,45)(H4,35,36,37);1H

InChI key

CTKOXUOHQOJPNV-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Substrates

A sensitive and specific fluorogenic substrate for factor Xa.

comparable product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assay of coagulation proteases using peptide chromogenic and fluorogenic substrates.
R Lottenberg et al.
Methods in enzymology, 80 Pt C, 341-361 (1981-01-01)
T Morita et al.
Journal of biochemistry, 82(5), 1495-1498 (1977-11-01)
Twenty peptide-4-methylcoumarin amides (MCA) were newly synthesized and tested as possible substrates for alpha-thrombin, factor Xa, kallikreins, urokinase, and plasmin. These fluorogenic peptides contained arginine-MCA as the carboxyl-terminus. Release of 7-amino-4-methylcoumarin was determined fluorometrically. Of these peptides, the following were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service