Skip to Content
Merck
All Photos(1)

Key Documents

AXYZTF200RS

Pipette filter tips for Zymark® automated liquid handling system

size 200 μL, pkg of 5x960tips/cs, clear, racked, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
size:
200 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc ZTF-200-R-S
Corning
Pricing and availability is not currently available.

sterility

sterile

manufacturer/tradename

Axygen Inc ZTF-200-R-S
Corning

packaging

pkg of 5x960tips/cs

size

200 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Products are available in China only.

Other Notes

Browse all pipette tips

Legal Information

Zymark is a registered trademark of Sotax Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service