Skip to Content
Merck
All Photos(1)

Documents

AXYPCR96LC480WNF

PCR microplate for Roche® 480 LightCycler®

size 96 wells, white, no sealing film

Synonym(s):

LightCycler Plate, PCR Microplate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

white
white polypropylene wells

sterility

non-sterile

packaging

case of 50 ea

manufacturer/tradename

Axygen Inc
Corning

technique(s)

PCR: suitable

size

96 wells

working volume

100 mL

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
LightCycler is a registered trademark of Roche
Roche is a registered trademark of Roche Molecular Systems, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service