Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR96HSACC

Corning® Axygen® 96 Well Automation Compatible PCR Microplate

Half Skirt, Color Waves polypropylene, non-sterile

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
Color Waves polypropylene
size:
96 well
sterility:
non-sterile
feature:
skirt
Pricing and availability is not currently available.

material

Color Waves polypropylene

sterility

non-sterile

feature

skirt

manufacturer/tradename

Corning PCR-96-HS-AC-C

size

96 well

General description

Axygen automation-compatible PCR microplate features a rigid frame with a small split that sustains heat transferring during and post thermal cycling without distorting its shape. The polypropylene plate also features raised rims around the wells to provide tight sealing to minimize heat evaporation. Compatible with most major thermal cyclers, sequencers, and automation platforms, the plate provides optimal performance for downstream applications, including high throughput screening.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service