Skip to Content
Merck
All Photos(1)

Documents

AV48526

Sigma-Aldrich

Anti-TRMT11 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-C6orf75, Anti-MDS024, Anti-TRM11, Anti-dJ187J11, Anti-dJ187J11.2, Anti-tRNA methyltransferase 11 homolog (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

human, mouse, horse, bovine, dog, rabbit, rat, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRMT11(60487)

General description

TRMT11-1 codes for tRNA Methyltransferase 11 homolog (S. cerevisiae). It may be involved in the processing of tRNA. Genetic variation in TRMT11 has been analyzed as a biomarker to predict the efficacy of androgen deprivation therapy in prostate cancer patients.
Rabbit Anti-TRMT11 antibody recognizes human, mouse, rat, chicken, canine, and bovine TRMT11.

Immunogen

Synthetic peptide directed towards the N terminal region of human TRMT11

Application

Rabbit Anti-TRMT11 antibody is suitable for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

TRMT11 belongs to the methyltransferase superfamily. It is a catalytic subunit of an S-adenosyl-L-methionine-dependent tRNA methyltransferase complex that mediates the methylation of the guanosine nucleotide at position 10 (m2G10) in tRNAs.

Sequence

Synthetic peptide located within the following region: IKIHTFNKTLTQEEKIKRIDALEFLPFEGKVNLKKPQHVFSVLEDYGLDP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manish Kohli et al.
Mayo Clinic proceedings, 87(3), 240-246 (2012-03-06)
To evaluate whether germline variations in genes involved in sex steroid biosynthesis and metabolic pathways predict time to treatment failure for patients with advanced prostate cancer undergoing androgen deprivation therapy (ADT), because there are few known clinical predictors of response.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service