Synthetic peptide directed towards the N terminal region of human SLC39A5
Application
Anti-SLC39A5 (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.
Biochem/physiol Actions
SLC39A5 (ZIP5) is a zinc uptake transporter that specifically transports Zn+2 ions. It is expressed in intestine, pancreas, liver and kidney. ZIP5 maintains zinc homeostasis and plays an important role in the uptake of dietary zinc across apical membrane of enterocytes in the intestine. Zinc transport by ZIP5 is important to protect the pancreatic acinar cells against zinc toxicity.
Sequence
Synthetic peptide located within the following region: HYLAQLFGLYGENGTLTAGGLARLLHSLGLGRVQGLRLGQHGPLTGRAAS
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
ZIP5 localizes to the baso-lateral membranes of intestinal enterocytes and pancreatic acinar cells and is internalized and degraded coordinately in these cell-types during periods of dietary zinc deficiency. These cell-types are thought to control zinc excretion from the body. The
The Journal of biological chemistry, 279(49), 51433-51441 (2004-08-24)
The mouse and human Zip5 proteins are members of the ZIP family of metal ion transporters. In this study, we present evidence that mouse Zip5 is a zinc uptake transporter that is specific for Zn(II) over other potential metal ion
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 25(2), 319-335 (2011-11-25)
Translation of the basolateral zinc transporter ZIP5 is repressed during zinc deficiency but Zip5 mRNA remains associated with polysomes and can be rapidly translated when zinc is repleted. Herein, we examined the mechanisms regulating translation of Zip5. The 3'-untranslated region
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.