Skip to Content
Merck
All Photos(1)

Documents

AV42559

Sigma-Aldrich

Anti-ANKRD2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ARPP, Anti-Ankyrin repeat domain 2 (stretch responsive muscle), Anti-MGC104314

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

49 kDa

species reactivity

guinea pig, horse, mouse, rabbit, human, dog, bovine, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANKRD2(26287)

General description

Ankyrin repeat domain 2 (stretch responsive muscle) (Ankrd2), a MARP mechanosensing protein, is believed to be involved in skeletal muscle hypertrophy through the regulation of cell signaling and gene expression within muscle cells.

Specificity

Anti-ANKRD2 antibody reacts with canine, human, mouse, and rat ankyrin repeat domain 2 (stretch responsive muscle) proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human ANKRD2

Application

Anti-ANKRD2 polyclonal antibody is used to tag Ankyrin repeat domain 2 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role Ankrd2 in muscle cell development.

Biochem/physiol Actions

ANKRD2 may play an important role in skeletal muscle hypertrophy.

Sequence

Synthetic peptide located within the following region: QEEENEKLRGDARQKLPMDLLVLEDEKHHGAQSAALQKVKGQERVRKTSL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service