Skip to Content
Merck
All Photos(1)

Key Documents

A6331

Sigma-Aldrich

N-Acetyl-S-geranylgeranyl-L-cysteine

≥95%, ethanol solution

Synonym(s):

AGGC, N-Acetyl-S-(3,7,11,15-tetramethyl-2E,6E,10E,14-hexadecatetraenyl)-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H41NO3S
CAS Number:
Molecular Weight:
435.66
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥95%

form

ethanol solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC\C(C)=C\CSC[C@H](NC(C)=O)C(O)=O

Biochem/physiol Actions

Synthetic substrate for the isoprenylated protein methyltransferase (S-adenosylmethionine-dependent methyltransferase). Functions as an inhibitor of methylation of endogenous isoprenylated proteins.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service