Skip to Content
Merck
All Photos(1)

Key Documents

A2558

Sigma-Aldrich

N-Acetylneuraminyl-fucosyllacto-N-neo-tetraose from human milk

Synonym(s):

α-NeuNAc-(2→6)-β-Gal-(1→4)-β-GlcNAc-(1→3)-β-Gal-(1→4)-(α-Fuc-[1→3])-Glc, SFLNnT, Sialyl-fucosyllacto-N-neo-tetraose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H72N2O33
CAS Number:
Molecular Weight:
1145.03
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

human milk

form

solid

storage temp.

−20°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)C(O)O[C@H](CO)[C@@H]2O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O[C@@H]4O[C@H](CO)[C@@H](O[C@@H]5O[C@H](CO[C@@]6(C[C@H](O)[C@@H](NC(C)=O)[C@@H](O6)[C@H](O)[C@H](O)CO)C(O)=O)[C@H](O)[C@H](O)[C@H]5O)[C@H](O)[C@H]4NC(C)=O)[C@H]3O)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C43H72N2O33/c1-10-21(54)26(59)28(61)39(69-10)77-36-30(63)37(65)70-17(8-49)33(36)75-41-31(64)35(24(57)15(6-47)71-41)76-38-20(45-12(3)51)25(58)32(16(7-48)72-38)74-40-29(62)27(60)23(56)18(73-40)9-68-43(42(66)67)4-13(52)19(44-11(2)50)34(78-43)22(55)14(53)5-46/h10,13-41,46-49,52-65H,4-9H2,1-3H3,(H,44,50)(H,45,51)(H,66,67)/t10-,13-,14+,15+,16+,17+,18+,19+,20+,21+,22+,23-,24-,25+,26+,27-,28-,29+,30+,31+,32+,33-,34+,35-,36+,37?,38-,39-,40-,41-,43+/m0/s1

InChI key

HSYHUIKGLHMONM-RDNIXPOZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Acetylneuraminyl-fucosyllacto-N-neo-tetraose is used as a reference material in the analysis of milk oligosaccharides.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service