Skip to Content
Merck
All Photos(1)

Documents

A2227

Sigma-Aldrich

Adrenocorticoptropic Hormone Fragment 22 - 39

≥95% (HPLC)

Synonym(s):

β-Cell-Tropin, ACTH (22-39), Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C90H125N19O32
Molecular Weight:
1985.06
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

sterility

non-sterile

Assay

≥95% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@@H]1CCCN1C(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@H](CO)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)CNC(=O)[C@H](CC(N)=O)NC(=O)[C@@H]3CCCN3C(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@@H](N)C(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc5ccccc5)C(O)=O

InChI

1S/C90H125N19O32/c1-44(2)36-57(82(132)99-55(28-32-70(118)119)80(130)106-62(90(140)141)39-50-18-12-9-13-19-50)102-85(135)64-20-14-34-108(64)88(138)60(37-49-16-10-8-11-17-49)104-76(126)48(7)95-78(128)53(26-30-68(114)115)97-75(125)47(6)96-84(134)63(43-110)107-81(131)56(29-33-71(120)121)100-83(133)59(41-72(122)123)101-79(129)54(27-31-69(116)117)98-74(124)46(5)94-67(113)42-93-77(127)58(40-66(91)112)103-86(136)65-21-15-35-109(65)89(139)61(105-87(137)73(92)45(3)4)38-51-22-24-52(111)25-23-51/h8-13,16-19,22-25,44-48,53-65,73,110-111H,14-15,20-21,26-43,92H2,1-7H3,(H2,91,112)(H,93,127)(H,94,113)(H,95,128)(H,96,134)(H,97,125)(H,98,124)(H,99,132)(H,100,133)(H,101,129)(H,102,135)(H,103,136)(H,104,126)(H,105,137)(H,106,130)(H,107,131)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,122,123)(H,140,141)/t46-,47-,48-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-,64-,65-,73-/m0/s1

InChI key

VNYTZTMGIJDKGF-SYGRCPSDSA-N

Gene Information

human ... POMC(5443)

Amino Acid Sequence

Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Celentano et al.
Journal of cellular physiology, 234(3), 2013-2020 (2018-09-22)
Synthetic corticosteroids are routinely administered during the treatment of several diseases, including malignancies. However, recent evidence suggests that corticosteroids may have tumor-promoting effects, particularly in epithelial neoplasms. Our aim was to assess the role of the recently characterized cancer-associated glucocorticoid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service