Skip to Content
Merck
All Photos(1)

Documents

A0733

Sigma-Aldrich

ABL2 (38-end), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ABLL, ARG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1035-1401 nmol/min·mg

mol wt

~132 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ABL2(27)

Biochem/physiol Actions

ABL2 (or ARG) is a nonreceptor cytoplasmic tyrosine kinase which is closely related to but distinct from ABL1. The similarity of ABL1 and ABL2 includes the tyrosine kinase domains and extends amino-terminal to include the SH2 and SH3 domains. ABL2 is involved in translocation with the ETV6 gene in human leukemia and has an altered expression in several human carcinomas. Two isoforms of ABL2 with different N-termini (1A and 1B) have been identified. The C-terminal domain of ABL2 contains two F-actin-binding sequences that perform a number of actions related to cell morphology and motility by interacting with actin filaments.

Physical form

Supplied in 50mM NaPhosphate pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1mM PMSF, 0.2 mM DTT, 25% glycerol.
.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank Griesinger et al.
British journal of haematology, 119(2), 454-458 (2002-10-31)
ETV6, a member of the Ets family of transcription factors, is frequently rearranged to various translocation partners in human leukaemias. We previously described a CD3+/TCRalpha/beta+ mature T-cell acute lymphoblastic leukaemia (T-ALL) cell line, MT-ALL, carrying a t(1;10;12)(q25; p13;p13) with cytokine-inducible
D Barilá et al.
Nature genetics, 18(3), 280-282 (1998-03-21)
The ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase (c-Abl) that has been implicated in processes of cell differentiation, cell division, cell adhesion and stress response. Alterations of ABL1 by chromosomal rearrangement or viral transduction can lead to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service