41494
0.1 M HEPES pH 7.0 − 30% PEG 6000 solution
Synonym(s):
PEG Grid Screening Solution 40/Kit-No 36436
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
form
liquid
pH
7.0±0.2 ( in neat)
density
1.06 g/mL at 20 °C
storage temp.
2-8°C
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service