Skip to Content
Merck
All Photos(1)

Key Documents

405M-1

Sigma-Aldrich

FOXA1 (2F83) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€142.00
0.1 ML
€189.00
0.5 ML
€411.00
1 ML
€614.00

€142.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€142.00
0.1 ML
€189.00
0.5 ML
€411.00
1 ML
€614.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€142.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

2F83, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (405M-14)
vial of 0.5 mL concentrate (405M-15)
bottle of 1.0 mL predilute (405M-17)
vial of 1.0 mL concentrate (405M-16)
bottle of 7.0 mL predilute (405M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1κ

control

breast carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... FOXA1(3169)

General description

Forkhead box A1 (FOXA1), also known as hepatocyte nuclear factor 3 alpha(HNF3a), is a transcription factor expressed in normal breast ductal epithelium and other epithelium in different organs, such as lung, pancreas, bladder, prostate, and colon. It has been reported to co-express with ER in breast carcinoma, predominantly in luminal subtype A.[1][2] FOXA1 expression has been identified in 42% - 75% of invasive carcinoma of the breast.[1][2] Of the FOXA1-positive breast invasive carcinomas, 83% were comprised of ER+/PR+/Her2+ luminal A subtype.[1] The results indicate that anti-FOXA1 is useful in the sub-classification of breast carcinoma.

Quality


IVD

IVD

IVD

RUO

Linkage

FOXA1 Positive Control Slides, Product No. 405S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Thorat et al.
Journal of clinical pathology, 61(3), 327-332 (2007-11-27)
Forkhead box A1 (FOXA1) is a forkhead family transcription factor expressed in breast cancer cells. It is essential for optimal expression of approximately 50% of oestrogen receptor (ER)-related genes. This study explored the FOXA1 relationship with luminal and basal breast
André Albergaria et al.
Breast cancer research : BCR, 11(3), R40-R40 (2009-06-25)
The expression of additional genes, other than oestrogen receptor (ER), may be important to the hormone-responsive phenotype of breast cancer. Microarray analyses have revealed that forkhead box A1 (FOXA1) and GATA binding protein 3 (GATA-3) are expressed in close association

Questions

  1. Does the FOXA1 (2F83) antibody only react to human cells, or does it also react to mouse cells?

    1 answer
    1. Cell Marque IVD antibodies are only tested for use on human tissue in a clinical setting with immunohistochemistry only. No other methods such as Fluorescence or Flow Cytometry or western blot or other species such as mouse, rabbit, monkey, etc. have been qualified to work with those antibodies.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service