31748
Sudan Orange G
≥97% (HPLC)
Synonym(s):
2,4-Dihydroxyazobenzene, 4-(Phenylazo)resorcinol, SOG
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5N=NC6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
214.22
Colour Index Number:
11920
Beilstein:
958430
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
Recommended Products
Assay
≥97% (HPLC)
mp
143-146 °C (lit.)
λmax
≤388 nm (lit.)
SMILES string
Oc1ccc(\N=N\c2ccccc2)c(O)c1
InChI
1S/C12H10N2O2/c15-10-6-7-11(12(16)8-10)14-13-9-4-2-1-3-5-9/h1-8,15-16H/b14-13+
InChI key
BPTKLSBRRJFNHJ-BUHFOSPRSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Daniel Kessler et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(50), 16555-16566 (2014-12-17)
Previous neuroimaging investigations in attention-deficit/hyperactivity disorder (ADHD) have separately identified distributed structural and functional deficits, but interconnections between these deficits have not been explored. To unite these modalities in a common model, we used joint independent component analysis, a multivariate
Ze Wang et al.
Drug and alcohol dependence, 152, 102-108 (2015-05-11)
The functional interconnections of the addicted brain may differ from the non-addicted population in important ways, but prioranalytic approaches were usually limited to the study of connections between a few number of selected brain regions. Recent approaches enable examination of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service