Skip to Content
Merck
All Photos(1)

Key Documents

02848

Sigma-Aldrich

Ethanol – 2-propanol mixture

BioUltra, for molecular biology, 19:1, A15 ISO1, ≥99.5% (GC)

Synonym(s):

Ethanol : 2-propanol, Ethyl alcohol:Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
6204567
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

product line

BioUltra

Assay

≥99.5% (GC)

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.001% free alkali (as NH3)
≤0.002% free acid (as CH3COOH)
<0.001% aldehyde (as CH3CHO)
<0.001% formaldehyde (HCHO)
<0.001% ketone (as CH3COCH3)
<0.2% water

refractive index

n20/D 1.362

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: ≤0.02
λ: 280 nm Amax: ≤0.01

format

mixture

SMILES string

CCO.CC(C)O

InChI

1S/C3H8O.C2H6O/c1-3(2)4;1-2-3/h3-4H,1-2H3;3H,2H2,1H3

InChI key

AINBZKYUNWUTRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Precipitation of nucleic acids
Sales restrictions may apply

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bumjin Jang et al.
ACS nano, 11(6), 6146-6154 (2017-06-08)
Conventional photocatalytic micromotors are limited to the use of specific wavelengths of light due to their narrow light absorption spectrum, which limits their effectiveness for applications in biomedicine and environmental remediation. We present a multiwavelength light-responsive Janus micromotor consisting of
Precipitation of nucleic acids.
D M Wallace
Methods in enzymology, 152, 41-48 (1987-01-01)
Oruç Köklükaya et al.
Carbohydrate polymers, 255, 117468-117468 (2021-01-14)
Layer-by-Layer (LbL) assembled nanocoatings are exploited to impart flame-retardant properties to cellulosic substrates. A model cellulose material can make it possible to investigate an optimal bilayer (BL) range for the deposition of coating while elucidating the main flame-retardant action thus
Xiangsheng Zhang et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 148, 111957-111957 (2021-01-01)
Plumbagin (PLB) is a naphthoquinone endowed with potential medicinal properties, including anticancer activities. We evaluated the effects of PLB on the viability, cell cycle, autophagy, and apoptosis of endometrial carcinoma Ishikawa cells. The proliferation of cells was significantly inhibited by
Noemí Dorival-García et al.
Journal of chromatography. A, 1513, 69-77 (2017-07-26)
Despite their advantages, there is concern that single-use systems used in biopharmaceutical manufacture might release potentially toxic substances during standard unit operations that negatively impact cell growth. Characterization of the extractables profile for single-use systems is necessary to know which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service