Skip to Content
Merck
All Photos(3)

Key Documents

T3812

Sigma-Aldrich

Tetracaine

meets USP testing specifications

Synonym(s):

4-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O2
CAS Number:
Molecular Weight:
264.36
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

CCCCNc1ccc(cc1)C(=O)OCCN(C)C

InChI

1S/C15H24N2O2/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3/h6-9,16H,4-5,10-12H2,1-3H3

InChI key

GKCBAIGFKIBETG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Topical ophthalmic anesthetic; used for spinal anesthesia

Biochem/physiol Actions

Blocks voltage-sensitive release of Ca2+ from sarcoplasmic reticulum.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Howard Bregman et al.
Journal of medicinal chemistry, 54(13), 4427-4445 (2011-06-04)
Clinical human genetic studies have recently identified the tetrodotoxin (TTX) sensitive neuronal voltage gated sodium channel Nav1.7 (SCN9A) as a critical mediator of pain sensitization. Herein, we report structure-activity relationships for a novel series of 2,4-diaminotriazines that inhibit hNav1.7. Optimization
Anna Taddio et al.
The Annals of pharmacotherapy, 36(4), 687-692 (2002-03-29)
To determine the relative efficacy of lidocaine-prilocaine and tetracaine for procedural pain in children. Systematic review was performed (MEDLINE1990-June 2001) of all studies comparing the efficacy of these 2 modalities of pain relief in children undergoing painful cutaneous procedures. Search
Lotta Salmela et al.
International journal of pharmaceutics, 472(1-2), 276-281 (2014-06-20)
We present a continuous-flow method that allows the release of drugs from submicron colloidal carriers to be estimated on a millisecond timescale. The technique is applied to the study of release of a model drug (tetracaine) from lipid emulsions, and
Ryan M Peters et al.
Journal of neurophysiology, 114(1), 264-273 (2015-05-01)
Galvanic vestibular stimulation (GVS) evokes a perception of rotation; however, very few quantitative data exist on the matter. We performed psychophysical experiments on virtual rotations experienced when binaural bipolar electrical stimulation is applied over the mastoids. We also performed analogous
Yveline Henchoz et al.
Journal of medicinal chemistry, 51(3), 396-399 (2008-01-15)
Accurate determinations of lipophilicity indices benefit from recent advances in chromatographic sciences such as the launch of ultraperformance liquid chromatography (UPLC). The fast strategy presented here emerges as a powerful method suitable for high-throughput log P measurements of therapeutic compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service