Skip to Content
Merck
All Photos(1)

Documents

QC1329

Supelco

Phenolics, Total

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

manufacturer/tradename

RTC QCI-1373

concentration

in acetone

technique(s)

UV/Vis spectroscopy: suitable

application(s)

environmental
food and beverages

format

matrix material

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weimin Peng et al.
Genetics, 179(1), 277-289 (2008-05-22)
Histone acetylation levels are regulated through the opposing activities of histone acetyltransferases (HATs) and deacetylases (HDACs). While much is known about gene-specific control of histone acetylation, little is understood about how total or cellular levels of histone acetylation are regulated.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service