PHR1978
Ibuprofen Related Compound J
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(2RS)-2-(4-Isobutyrylphenyl)propionic acid, (2-(4-Isobutyrylphenyl)propanoic acid), α-Methyl-4-(2-methyl-1-oxopropyl)benzeneacetic acid, (2RS)-2-[4-(2-Methylpropanoyl)phenyl]propanoic acid, 1-Oxoibuprofen
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1335596
CofA
current certificate can be downloaded
packaging
pkg of 50 mg
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-30°C
SMILES string
OC(C(C)C1=CC=C(C(C(C)C)=O)C=C1)=O
InChI
1S/C13H16O3/c1-8(2)12(14)11-6-4-10(5-7-11)9(3)13(15)16/h4-9H,1-3H3,(H,15,16)
InChI key
RCINGEKPKJQCMO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Prediction of Potential Toxicity and Side Effect Protein Targets: Research exploring the use of Ibuprofen Related Compound J as part of a ligand-protein inverse docking approach aimed at predicting potential toxicity and side effect protein targets. This innovative method enhances drug safety profiles by identifying off-target interactions early in the drug development process, serving as a critical tool for pharmaceutical research and ensuring the development of safer therapeutic agents (Chen and Ung, 2001).
Analysis Note
Other Notes
Footnote
Recommended products
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service