Skip to Content
Merck
All Photos(1)

Key Documents

PE1621

Supelco

Trihalomethanes DE-PT

Proficiency Testing Material

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
77101502

packaging

ampule of 2 mL

application(s)

environmental

storage temp.

2-8°C

Other Notes

Proficiency Testing Schedule (2018-2019)

Drinking Water
WS18-1January 17 - March 2, 2018
WS18-2March 28 - May 11, 2018
WS18-3July 11 - August 24, 2018
WS18-4September 26 - November 9, 2018

WS19-1January 16 - March 1, 2019
WS19-2March 27 - May 10, 2019
WS19-3 July 10 - August 23, 2019
WS19-4 September 25 - November 8, 2019

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

    Bromodichlormethane .4-20 μg/L
    Dibromochlormethane .4-20 μg/L
    Tribrommethane .4-20 μg/L
    Trichloromethane .4-20 μg/L

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service