Skip to Content
Merck
All Photos(1)

Key Documents

NIST3389

Ginsenosides Calibration Solutions

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

packaging

pkg of 5 × 1 mL

manufacturer/tradename

NIST®

concentration

in methanol

application(s)

agriculture

format

multi-component solution

storage temp.

−20°C

General description

Each unit of SRM 3389 contains five ampoules of ~1 milliliter of ginsenoside with:
1.four ampoules comprising a mixture of six-component ginsenosides Rb1, Rb2, Rc, Rd, Re, and Rg1 in methanol, and
2.one ampoule with ginsenoside Rf in methanol
all of which can be stored at -20 °C or below.
For more information, please refer to the SDS and COA.
SRM 3389_cert
SRM 3389_SDS

Application
(1) Certified values provided by SRM 3389 are intended to produce metrologically traceable calibrants used in the determination of Rb1, Rb2, Rc, Rd, Re, and Rg1 ginsenosides in natural matrix samples.
(2) Non-certified value provided by this SRM is intended for the harmonization of ginsenoside Rf measurements in natural matrix samples.

Features and Benefits

  • Certifiedvalues for various constituents of ginsenosides are provided, values aremetrologically traceable to SI unit and expressed as Mass Fraction (mg/g).
  • Non-certified mass fraction values are provided for ginsenoside Rf and values are idealfor method harmonization, method development, and process control.
  • The non-certified values are metrologically traceable to the materials and methods employed in its determination.
  • NIST certificate is provided with expiration details, safety, source, instructions for storage and use.

Other Notes

Certified for the analytes listed below.
Ginsenoside Rg1 (C42H72O14), Ginsenoside Re (C48H82O18), Ginsenoside Rb1 (C54H92O23), Ginsenoside Rc (C53H90O22), Ginsenoside Rb2 (C53H90O22), Ginsenoside Rd (C48H82O18)

Reference/Informational value is provided for the following analytes.
Ginsenoside Rf (C42H72O14)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service