Skip to Content
Merck
All Photos(2)

Documents

NIST2720

Sulfur in di-n-butyl sulfide

NIST® SRM® 2720

Synonym(s):

Dibutyl sulfide, 1,1′-Thiobisbutane, Butyl sulfide, NSC 8460

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3S(CH2)3CH3
CAS Number:
Molecular Weight:
146.29
Beilstein:
1732829
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

vapor density

5.07 (vs air)

vapor pressure

5.17 mmHg ( 37.7 °C)

packaging

pkg of 5 x 4.5 mL

manufacturer/tradename

NIST®

technique(s)

XRF: suitable

refractive index

n20/D 1.452 (lit.)

bp

188-189 °C (lit.)

mp

−76 °C (lit.)

density

0.838 g/mL at 25 °C (lit.)

application(s)

petroleum

format

matrix material

SMILES string

CCCCSCCCC

InChI

1S/C8H18S/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3

InChI key

HTIRHQRTDBPHNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Sulfur (S)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

141.8 °F

Flash Point(C)

61 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service