NIST2191A
Sodium bicarbonate
NIST® SRM® 2191a, pD Standard
Synonym(s):
Sodium hydrogen carbonate
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
crystals
packaging
pkg of 30 g
manufacturer/tradename
NIST®
pH
8.3(0.84% solution)
pKa (25 °C)
(1) 6.37, (2) 10.25 (carbonic acid)
density
2.16 g/mL at 25 °C (lit.)
application(s)
general analytical
SMILES string
[Na+].OC([O-])=O
InChI
1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1
InChI key
UIIMBOGNXHQVGW-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- SRM 2191A certified values are correspond to log (1/aD).
- The NIST certificate is provided with storage and stability instructions.
Other Notes
Legal Information
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service